Order Entry
United Kingdom
ContactUsLinkComponent
D-(+)-Sucrose, white crystalline powder
D-(+)-Sucrose, white crystalline powder
Supplier:  MP Biomedicals
Certificates
undefined
D-(+)-Sucrose, white crystalline powder
Supplier:  MP Biomedicals

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard
  • Optical Rotation:
    66 + 3° (C=2 water)
  • Melting Point:
    170 - 190 °C

Specification Test Results

Frequently Bought Together