Reactive red 4, powder C.I. 18105
: tetrapotassium 5-benzamido-3-{2-[5-({4-chloro-6-[(4-sulphonatophenyl)amino]-1,3,5-triazin-2-yl}amino)-2-sulphonatophenyl]diazen-1-yl}-4-hydroxynaphthalene-2,7-disulphonate, Procion Reactive Red 4, Cibacron Brilliant Red 3B-A
:17681-50-4
:
:C₃₂H₁₉ClN₈O₁₄S₄Na₄
:MFCD00009690
:995.2 g/mol
:Ambient
- Environmentally Preferable:
- Identity Test:Passes
- λmax (ethanol):282 ±2 nm
Frequently Bought Together