Benzalkonium chloride ≥80%, viscous solution
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
CAS Number:8001-54-5
Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
MDL Number:MFCD00145757
Storage Temperature:Ambient
UN:2923
ADR:8 III
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- Concentration:80% Solution
Specification Test Results
Frequently Bought Together