Order Entry
United Kingdom
ContactUsLinkComponent
DNA ladder, 100 bp
DNA ladder, 100 bp
Catalog # A5191.0025
Supplier:  PanReac AppliChem
CAS Number:  
undefined
DNA ladder, 100 bp
Catalog # A5191.0025
Supplier:  PanReac AppliChem
CAS Number:  
Specifications
  • Pk:
    0,25 mg
  • Description:
    DNA ladder, 100 bp
  • Environmentally Preferable:
Specifications
About this item
This DNA ladder is ideal for determining the size of double-stranded DNA ranging from 100 to 1,000 base pairs. The ladder consists of ten fragments that range in size from 100 to 1,000 bp in 100 bp increments. The 500 bp fragment is present at increased intensity to allow easy identification. All fragments are blunt-ended.
The marker is supplied in storage buffer (10 mM Tris · HCl (pH 7,8), 10 mM NaCl, 1 mM EDTA) in a concentration of 0,2 mg/ml. The recommended amount of size marker to load on the gel is 0,4 to 0,6 μg (2 to 3 μl) per lane.