Order Entry
United Kingdom
ContactUsLinkComponent
Coating thickness gauges, PosiTest DFT
Coating thickness gauges, PosiTest DFT
Supplier:  Industrial Physics
Certificates
undefined
Coating thickness gauges, PosiTest DFT
Supplier:  Industrial Physics
Specifications
  • Catalog No:
  • SIMELD6011
  • SIMELD6012
  • Model:
  • PosiTest DFT Fe
  • PosiTest DFT Fe/NFe
  • Accuracy:
  • ±2 μm +3 %
  • ±2 μm +3 %
  • W×D×H:
  • 100×38×23 mm
  • 100×38×23 mm
  • Weight:
  • 70 g
  • 70 g