Order Entry
United Kingdom
ContactUsLinkComponent
Anti-FKBP6 Rabbit Polyclonal Antibody
Anti-FKBP6 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30078
Supplier:  Abnova
undefined
Anti-FKBP6 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30078
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    FKBP6
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    8468
  • Sequence:
    FCALSAEQQDQFPLQKVLKVAATEREFGNYLFRQNRFYDAKVRYKRALLL
  • Size:
    100 μL
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of human FKBP6.

Type: Primary
Antigen: FKBP6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human