Order Entry
United Kingdom
ContactUsLinkComponent
Impact tester acc. to ASTM G14
Impact tester acc. to ASTM G14
Catalog # SIMESP1895
Supplier:  Industrial Physics
undefined
Impact tester acc. to ASTM G14
Catalog # SIMESP1895
Supplier:  Industrial Physics
Specifications
  • Description:
    Impact test acc. to ASTM G14
  • Seq:
    03
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Impact tester used to determine the impact resistivity and flexibility of materials.
  • Spirit level built-in for correct positioning
  • Dual scale instrument equipped with a special guidance
  • Distance between each impact is always according to the standards
Ordering information: Includes base plate, guide tube, release collar, punch (Ø = 16 mm) and V-notch vise with spring clamp to hold the pipe.