Specifications
- Antibody type:Primary
- Antigen name:breakpoint cluster region
- Antigen symbol:BCR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Lyophilized
- Gene ID:613
- Antigen synonyms:PHL|D22S11|BCR1|ALL|CML|D22S662
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:E.coli-derived human Bcr recombinant protein (Position: M1-A100). Human Bcr shares 94% amino acid (aa) sequence identity with mouse Bcr.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
The breakpoint cluster region protein (BCR) is a protein that in humans is encoded by the BCR gene. A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene.
1. "Entrez Gene: Breakpoint cluster region".
2. Puil L, Liu J, Gish G, Mbamalu G, Bowtell D, Pelicci PG, Arlinghaus R, Pawson T (February 1994). "Bcr-Abl oncoproteins bind directly to activators of the Ras signalling pathway". EMBO J. 13 (4): 764–73.
3. Pendergast AM, Muller AJ, Havlik MH, Maru Y, Witte ON (July 1991). "BCR sequences essential for transformation by the BCR-ABL oncogene bind to the ABL SH2 regulatory domain in a non-phosphotyrosine-dependent manner".Cell 66 (1): 161–71.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.