Cut resistant gloves, Check & Go Amber PU 3
Supplier: HONEYWELL SAFETY
Certificates
About this item
These polyamide/HPPE fibres/elastane knitted gloves with PU coating on palm and fingertips provide cut and puncture resistance. Ideal for use when handling sharp parts in a dry or greasy environment.
- Knitting technique helps produce a glove that fits the hands snugly, with good dexterity
- Tight knit (gauge 13) gives the glove perfect support, limits penetration of dirt and offers good sense of touch
- Ventilated back optimises breathability
- PU coating of the palm and fingertips provides outstanding abrasion resistance
- Colour-coding on top of the hand and coating colour helps safety manager and workers to visually recognise the level of cut protection
Typical application areas include: Automotive and part manufacturing, aviation building and construction, catering, energy or electricity, glass industries, metal and paper industries, logistics and maintenance.
Caution: Do not expose this glove to the heat.
Certifications: EN 388 (4342); Cat. II
Caution: Please note: Do not expose this glove to heat.
Specifications
- Catalog No:
- 112-4643
- 112-4644
- 112-4647
- 112-4645
- 112-4646
- Model:
- Check & Go Amber PU 3
- Check & Go Amber PU 3
- Check & Go Amber PU 3
- Check & Go Amber PU 3
- Check & Go Amber PU 3
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide/elastane/HPPE fibres
- Polyamide/elastane/HPPE fibres
- Polyamide/elastane/HPPE fibres
- Polyamide/elastane/HPPE fibres
- Polyamide/elastane/HPPE fibres
- Finish:
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (4342)
- EN 388 (4342)
- EN 388 (4342)
- EN 388 (4342)
- EN 388 (4342)