2-Pyrimidinecarboxylic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Pyrimidine-2-carboxylic acid, 2-Carboxypyrimidine
CAS Number:31519-62-7
:
Formula:C₅H₄N₂O₂
MDL Number:MFCD00856161
Molecular Weight:124.1 g/mol
Melting Pt:190…195 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥94.0 %
Specification Test Results
Frequently Bought Together