tert-Butyl-3-aminoazetidine-1-carboxylate 95%
: 3-Amino-1-Boc-azetidine, tert-Butyl 3-aminoazetidine-1-carboxylate, 1-BOC-3-aminoazetidine
:193269-78-2
:
:C₈H₁₆N₂O₂
:MFCD01861753
:172.23 g/mol
:2735
:GHS05
:Ambient
- Environmentally Preferable:
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥94.0 %
Frequently Bought Together