Perfluorokerosene 50% in PFK low boiling, high boiling for mass spectrometry
Catalog # ACRO450130010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Suitability:(for mass analysis) Passes test
Specification Test Results
About this item
Synonyms: PFK
:
Boiling Pt:180 °C
UN:1993
:GHS02
Storage Temperature:Ambient
MDL Number:MFCD00132835
Density:1.89 g/cm³
Flash Pt:46 °C
ADR:3 III