Doxylamine succinate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:562-10-7
:
Formula:C₂₁H₂₈N₂O₅
MDL Number:MFCD00056168
Molecular Weight:388.464 g/mol
Melting Pt:103…108 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:103 to 108 °C
- Titration with HClO4:≥97.5 %
Specification Test Results
Frequently Bought Together