Order Entry
United Kingdom
ContactUsLinkComponent
 
Sodium molybdate(VI) dihydrate ACS
 
undefined
Sodium molybdate(VI) dihydrate ACS
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White Solid
  • Titration Complexometric:
    99.5 to 103.0 %
  • pH:
    7.0 to 10.5 (5% solution at 25.0°C)
  • Chloride (Cl):
    ≤0.005 %
  • Phosphate (PO4):
    ≤0.0005 %
  • Sulfate (SO4):
    ≤0.015 %
  • Ammonium (NH4):
    ≤0.001 %
  • Iron (Fe):
    ≤0.001 %
  • Heavy metals (as Pb):
    ≤5 ppm
  • Insoluble matter:
    ≤0.005 % (in water)

 

 

Frequently Bought Together