tri-Sodium citrate dihydrate ACS
: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate
:6132-04-3
:
:Na₃C₆H₅O₇·2H₂O
:MFCD00150031
:294.1 g/mol
:>300 °C
:13,08675
:Ambient
- Environmentally Preferable:
- Appearance:White Crystalline powder or crystals
- pH:7.0 to 9.0 (5 % in water at 25.0°C)
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤0.003 %
- Sulfate (SO4):≤0.005 %
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤5 ppm
- Heavy metals (ICP-OES):≤5 ppm
- Insoluble matter:≤0.005 %
- Ammonia (NH3):≤0.003 %
Frequently Bought Together