Order Entry
United Kingdom
ContactUsLinkComponent
Heptane (mixture of isomers), Technical Grade
Heptane (mixture of isomers), Technical Grade
Catalog # ACRO444270050
CAS Number:  
undefined
Heptane (mixture of isomers), Technical Grade
Catalog # ACRO444270050
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Boiling point:
    94 to 100°C
  • Density:
    0.685 to 0.730 kg/l (15°C)

Specification Test Results

About this item

 :
 :100.2 g/mol
UN:1206
 :GHS07
 :GHS08
 :GHS09
 :GHS02
Flash Pt:–4 °C
Storage Temperature:Ambient
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
Boiling Pt:98 °C (760 mmHg)
Melting Pt:–91 °C
Density:0.683 g/cm³
ADR:3 II
Merck Index:13,04679