Ethylene glycol, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
CAS Number:107-21-1
:
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885
Boiling Pt:196…198 °C (760 mmHg)
Melting Pt:–13 °C
Density:1.113 g/cm³
Molecular Weight:62.07 g/mol
Flash Pt:116 °C
Merck Index:13,03832
:GHS07
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Viscous liquid
- Refractive index:1.4300 to 1.4320 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together