Order Entry
United Kingdom
ContactUsLinkComponent
2-Chloro-4-fluoropyridine 97%
2-Chloro-4-fluoropyridine 97%
Certificates
undefined
2-Chloro-4-fluoropyridine 97%
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %