Potassium nitrate 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Nitrate of Potash
CAS Number:7757-79-1
:
Formula:KNO₃
MDL Number:MFCD00011409
Boiling Pt:400 °C (760 mmHg)
Molecular Weight:101.1 g/mol
Melting Pt:334 °C
ADR:5.1 III
Merck Index:13,07733
UN:1486
:GHS03
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder, crystals and chunks
- Titration after Ion exchange:99.0 to 100.5 %
- Chloride (Cl):≤0.002 %
- Sulfate (SO4):≤0.005 %
- Iron (Fe):≤0.001 %
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.1 % (in water)
Specification Test Results
Frequently Bought Together