trans-β-Styrylboronic acid pinacol ester 97%
: E-Phenylethenylboronic acid, pinacol ester, Trans-2-Phenylvinylboronic acid pinacol ester, trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene
:83947-56-2
:
:C₁₄H₁₉BO₂
:MFCD03453666
:89…93 °C (.1 mmHg)
:230.12 g/mol
:>110 °C
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Colorless to light yellow Liquid or low melting solid
- Infrared spectrum:Conforms
- GC:≥94.0 %
Frequently Bought Together