Methyl-2-aminoisonicotinate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Methyl 2-aminopyridine-4-carboxylate, Methyl 2-aminoisonicotinate
CAS Number:6937-03-7
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD04039316
Molecular Weight:152.15 g/mol
Melting Pt:145…148 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:144 to 148 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together