(1S,4R)-(+)-4-Acetoxy-2-cyclopentene-1-ol 99% stabilised
: (1S,4R)-cis-4-Acetoxy-2-cyclopenten-1-ol, (1R,4S)-4-hydroxycyclopent-2-enyl acetate, (1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate
:60410-16-4
:MFCD00210003
:C₇H₁₀O₃
:MFCD00078933
:142.154 g/mol
:43 °C
:Refrigerator
- Environmentally Preferable:
- Appearance:White to light yellow Solid
- Infrared spectrum:Conforms
- Melting point:49 to 53 °C
- GC:≥98.5 %
- Specific optical rotation:+67° to +69° (20 °C, 589 nm) (c=2.3, CHCl3)
Frequently Bought Together