Methylamine 33% (w/w) in ethanol absolute, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Aminomethane
CAS Number:74-89-5
:
Formula:CH₅N
MDL Number:MFCD00008104
Boiling Pt:48
Melting Pt:-92.5
Density:0.756 g/cm³
Molecular Weight:31.06 g/mol
Flash Pt:–23 °C
Storage Temperature:Ambient
ADR:3 II
Merck Index:12,06095
UN:2924
:GHS07
:GHS05
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Infrared spectrum:Conforms
- Water:≤1.0 %
- Titration with HClO4:31.0 to 35.0 %
Specification Test Results
Related Information
Frequently Bought Together