Order Entry
United Kingdom
ContactUsLinkComponent
Soda lime, granular, pure indicator, with indicator
Soda lime, granular, pure indicator, with indicator
Certificates
undefined
Soda lime, granular, pure indicator, with indicator
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White to beige to light grey Granules
  • Loss on drying:
    ≤20 % (10 g, 200°C, 2 h)
  • CO2 absorption capacity:
    ≥19.0 %