Bis(tri-o-tolylphosphine)palladium(II) dichloride 95%
: trans-Dichlorobis(tri-o-tolylphosphine)palladium(II), Bis[tri(o-tolyl)phosphine]palladium(II) chloride, Dichlorobis(tri-o-tolylphosphine)palladium(II), Dichlorobis(tri-o-tolylphsophine)palladium
:40691-33-6
:
:C₄₂H₄₂Cl₂P₂Pd
:MFCD00274659
:786.07 g/mol
:280 °C
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Yellow to orange Crystalline powder
- Infrared spectrum:Conforms
- Elemental Analysis:≥94 % (ex C)
Frequently Bought Together