Order Entry
United Kingdom
ContactUsLinkComponent
2-Amino-4,6-dimethylpyridine 99%
2-Amino-4,6-dimethylpyridine 99%
Catalog # ACRO103580050
CAS Number:  
undefined
2-Amino-4,6-dimethylpyridine 99%
Catalog # ACRO103580050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to brown Crystalline powder or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    64 to 70 °C
  • GC:
    ≥98.5 %
  • Heavy metals (as Pb):
    ≤2 ppm

Specification Test Results

About this item

 :
 :122.1700 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:5407-87-4
Formula:C₇H₁₀N₂
MDL Number:MFCD00006322
Boiling Pt:235 °C (760 mmHg)
Melting Pt:65…69.5 °C
Flash Pt:121 °C