2-Chloronicotinoyl chloride 97%
:49609-84-9
:
:C₆H₃Cl₂NO
:MFCD00051677
:89…91 °C (1 mmHg)
:176.00 g/mol
:38…39 °C
:>110 °C
:Ambient
:3261
:8 II
:GHS05
- Environmentally Preferable:
- Appearance:White to yellow to light beige Low melting solid
- Infrared spectrum:Conforms
- Melting point:39 to 44 °C
- GC:≥96.0 % (derivatized)
- NMR (1H-NMR):Consistent with structure
Frequently Bought Together