Ethyl hydrogen malonate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Mono-Ethyl malonate, 3-Ethoxy-3-oxopropanoic acid
CAS Number:1071-46-1
:
Formula:C₅H₈O₄
MDL Number:MFCD00020490
Boiling Pt:106.5 °C (3 mmHg)
Density:1.119 g/cm³
Molecular Weight:132.115 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Titration with NaOH:≥94 %
Specification Test Results
Frequently Bought Together