Diethyl ether ≥99% stabilised for HPLC
: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
:60-29-7
:
:(CH₃CH₂)₂O
:MFCD00011646
:35
:0.714 g/cm³
:Stabilised with ethanol
:74.12 g/mol
:–116 °C
:–45 °C
:3 I
:12,03852
:1155
:GHS07
:GHS02
:Ambient
- Environmentally Preferable:
- Appearance:Clear liquid
- Water:≤0.02 %
- GC:≥99 %
- UV:at 210 nm A: ≤0.3
- Color scale:≤10 APHA
- Stabilizer:2 % Ethanol
- Wavelength at 1 A.U.:≤205 nm
- Residue after evaporation:≤2 ppm
- Acidity/alkalinity:≤0.0002 meq/g
Frequently Bought Together