Copper(II) carbonate basic Cu 54-56%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
CAS Number:12069-69-1
:
Formula:CuCO₃·Cu(OH)₂
MDL Number:MFCD00010976
Molecular Weight:221.12 g/mol
Melting Pt:200
ADR:9 III
Merck Index:13,02658
UN:3077
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Green Crystalline powder
- Assay:54 to 56 % (Cu)
Specification Test Results
Frequently Bought Together