Order Entry
United Kingdom
ContactUsLinkComponent
 
Chromium(III) chloride, anhydrous 99%
 
undefined
Chromium(III) chloride, anhydrous 99%
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Purple Glistening powder or flakes
  • Assay:
    ≥98.5 %

 

 

Frequently Bought Together