Order Entry
United Kingdom
ContactUsLinkComponent
Nickel aluminium alloy according to Raney (Al-Ni) 50/50, powder
Nickel aluminium alloy according to Raney (Al-Ni) 50/50, powder
Certificates
undefined
Nickel aluminium alloy according to Raney (Al-Ni) 50/50, powder
Certificates
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Grey to grey-black Powder
  • Aluminium (Al):
    47.0 to 52.0 %
  • Nickel (Ni):
    48.0 to 53.0 %