Order Entry
United Kingdom
ContactUsLinkComponent
3-Chloropropylamine hydrochloride 98%
3-Chloropropylamine hydrochloride 98%
Certificates
undefined
3-Chloropropylamine hydrochloride 98%
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Off-white to yellow to light beige Crystalline powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥97.5 % (Total Chloride)
  • Water:
    ≤0.5 %