Phenol saturated solution in water stabilised for molecular biology DNase-, RNase-, protease-, oxides-free
Catalog # ACRO327105000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to orange Solution
- Water:25 to 35 %
- Stabilizer:0.08 to 0.12 % hydroxyquinoline
- Dnase,Rnase,Protease activity:Not detected
- Iron (Fe):≤1 ppm
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
About this item
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
:
:94.11 g/mol
UN:2821
:GHS06
:GHS08
:GHS05
:GHS09
Storage Temperature:Refrigerator
CAS Number:108-95-2
Formula:C₆H₅OH
MDL Number:MFCD00212506
Boiling Pt:181.7
Melting Pt:40.8
Flash Pt:79 °C
ADR:6.1 III
Merck Index:12,07390