Transfer pipettes, Samco™ Extra Long
Supplier: SAMCO
Certificates
About this item
These extra long PE pipettes are ideal for safe and easy sampling from large containers.
- Excellent for tissue culture applications
- General purpose pipettes are ideal for routine sampling of serum and plasma
- Well suited for blood culture and urine analysis
Perfect for tissue culture applications, these pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, autoclaving and prevents cross-contamination.
Specifications
- Catalog No:
- SAMC262-20S
- SAMC263
- SAMC262-10S
- SAMC262-1S
- SAMC263-1S
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)