Anti-FGF1 Antibody
BSBTPB9944
- Antibody type:Primary
- Antigen name:Fibroblast growth factor 1
- Antigen symbol:FGF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2246
- Antigen synonyms:AFGF|ECGFA|ECGF-beta|HBGF-1|FGF-alpha|HBGF1|FGFA|GLIO703|FGF-1|ECGF|ECGFB
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Fibroblast growth factor 1(FGF1) detection. Tested with WB, IHC-P, ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: FGF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat