Anti-CD9 Antibody
Catalog # BSBTPB9930
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:CD9 molecule
- Antigen symbol:CD9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:12527
- Antigen synonyms:Tspan29
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for CD9 antigen(CD9) detection. Tested with WB, IHC-P in Mouse.
Type: Primary
Antigen: CD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse