Anti-VIP Antibody
Catalog # BSBTRP1108
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Vasoactive intestinal peptide
- Antigen symbol:VIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7432
- Antigen synonyms:PHM27
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for VIP peptides(VIP) detection. Tested with IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: VIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat