Order Entry
United Kingdom
ContactUsLinkComponent
VWR®, Buffers, ammonia- ammonium chloride and ammonium chloride-ammonium hydroxide USP Test solutions (TS)
VWR®, Buffers, ammonia- ammonium chloride and ammonium chloride-ammonium hydroxide USP Test solutions (TS)
Catalog # 85326.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Buffers, ammonia- ammonium chloride and ammonium chloride-ammonium hydroxide USP Test solutions (TS)
Catalog # 85326.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Ammonia-ammonium chloride TS
  • Seq:
    01
  • Environmentally Preferable:
  • pH value:
    10,0

Specifications

About this item

Ready to use test solutions (TS).

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available