Copper(II) sulphate 0.5 mol/l USP Test Solution (TS)
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Cupric sulphate
CAS Number:7758-98-7
:
Formula:CuSO₄
MDL Number:MFCD00010981
Molecular Weight:159.61 g/mol
Melting Pt:200
ADR:9 III
Merck Index:12,02722
Storage Temperature:Ambient
EINECS:0
UN:3082
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together