Zinc acetate dihydrate 98.0-102.0%, crystals USP
Catalog # SPCMZI200-12KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly pail
- Pk:12 kg
- Seq:4187
- Environmentally Preferable:
Specifications
Specification Test Results
- ASSAY [Zn(CH3COO)2.2H2O]:98.0-102.0 %
- pH OF A 5% SOLUTION @ 25°C:6.0-8.0
- INSOLUBLE MATTER:0.005 %
- ARSENIC (As):3 ppm
- LEAD (Pb):0.002 %
- CHLORIDE (Cl):0.005 %
- SULFATES (SO4):0.010 %
- ALKALIES AND ALKALINE EARTHS:0.2 %
- ELEMENTAL IMPURITIES:AS REPORTED
- IDENTIFICATION:TO PASS TEST
- RESIDUAL SOLVENTS:TO PASS TEST
Specification Test Results
About this item
Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate
:
:219.51 g/mol
EINECS:209-170-2
UN:3077
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:5970-45-6
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961
Melting Pt:237
Density:1.718 g/cm³ (20 °C)
ADR:9 III
Merck Index:12,10256