Sodium stearate, powder NF
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Stearate sodium, Stearic acid sodium salt
CAS Number:822-16-2
:
Formula:C₁₈H₃₅NaO₂
MDL Number:MFCD00036404
Density:1.103 g/cm³ (20 °C)
Molecular Weight:306.46 g/mol
Melting Pt:288
Storage Temperature:Ambient
EINECS:212-490-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:90% Min.
Specification Test Results
Frequently Bought Together