L(+)-Potassium hydrogen tartrate USP
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Potassium hydrogen L-tartrate, Potassium acid tartrate, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Potassium Bitartrate, (R,R)-(+)-Tartaric acid potassium salt, L(+)-Tartaric acid potassium salt
CAS Number:868-14-4
:
Formula:KOOC(CHOH)₂COOH
MDL Number:MFCD00065392
Density:1.943 g/cm³ (20 °C)
Molecular Weight:188.18 g/mol
Melting Pt:250
Merck Index:13,07697
EINECS:212-769-1
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99% Min.
Specification Test Results
Frequently Bought Together