Ampicillin 100 mg/ml in aqueous solution ready-to-use
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69-53-4
:
Formula:C₁₆H₁₉N₃O₄S
MDL Number:MFCD00005175
Molecular Weight:349.4 g/mol
:Filtered through a 0.2 µm filter.
EINECS:200-708-1
:GHS08
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together