Phosphate, 0,5M buffer solution, pH 7,5
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together