Order Entry
United Kingdom
ContactUsLinkComponent
Leu-Enkephalin
Leu-Enkephalin
Catalog # J61408.MB
CAS Number:  
Leu-Enkephalin
Catalog # J61408.MB
CAS Number:  

Specifications

  • Pk:
    25 mg
  • CAS No.:
    81678-16-2
  • Source:
  • Species:
  • Environmentally Preferable:
  • Storage conditions:
    -30 °C to -10 °C
  • Form:
    Lyophilized powder
  • Protein/peptide name:
    Leu-Enkephalin
  • Molecular weight:
    555.70
  • Molecular formula:
    C₂₈H₃₇N₅O₇

Specifications

About this item

CAS No.: 81678-16-2 Molecular Formula: C28H37N5O7 Formula Weight: 555.70 Storage Temperature: -30°C to -10°C Physical Form: Lyophilized powder Appearance: White MDL No.: MFCD00076406

Applications: Endogenous opioid neurotransmitter that is an agonist at mu- and delta-opioid receptors