DNAzol® DNA isolation reagent
Applications: Complete and ready to use reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin
Physical Form: Liquid Appearance: Blue-green MDL No.: MFCD01093425
Applications: Complete and ready to use reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable:
Frequently Bought Together