Order Entry
United Kingdom
ContactUsLinkComponent
6-Boc-1-oxa-6-azaspiro[3.3]heptane 96%
6-Boc-1-oxa-6-azaspiro[3.3]heptane 96%
Catalog # H57040.MD
CAS Number:  
6-Boc-1-oxa-6-azaspiro[3.3]heptane 96%
Catalog # H57040.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

CAS Number:1223573-41-8
 :
Formula:C₁₀H₁₇NO₃
 :199.25 g/mol
 :GHS07
Storage Temperature:Ambient