Order Entry
United Kingdom
ContactUsLinkComponent
Methyl-2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate 97%
Methyl-2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate 97%
Catalog # H31668.03
CAS Number:  
Methyl-2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate 97%
Catalog # H31668.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate
 :
 :206.24 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:59604-96-5
Formula:C₁₂H₁₄O₃
MDL Number:MFCD09258894
Melting Pt:40…43 °C