Quercetin dihydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate
CAS Number:6151-25-3
:
Formula:C₁₅H₁₄O₉
MDL Number:MFCD00149487
Molecular Weight:338.268 g/mol
Melting Pt:310…315 °C
EINECS:204-187-1
ADR:6.1 III
:GHS06
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together